Check Order Status
Dichloromethane, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™
Element Search
Structure Search
Dichloromethane, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
5 L
Packaging:
View all
Amber Glass Bottle
Catalog number
FKCD185017
also known as FKCD/1850/17
Price (MYR)
152.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
152.00
EA
Add to cart
Specifications
Melting Point
-97°C
CAS
75-09-2
Color
Colorless
Boiling Point
39°C
Quantity
2.5 L
View more