▸
Specialty Lab Glassware
▸
Distillation Labware
▸
Distillation Columns, Vacuum Jacketed
Actual product may vary
Distillation Columns, Vacuum Jacketed
Vacuum jacketed distilling column for better performance
Have Questions?
Change view
Catalog Number
Standard Taper Joint
Height (Metric)
FKCC164250
also known as FKCC/1642/50
24/40
Jacket: 250mm
FGCC164250
also known as FGCC/1642/50
24/40
Jacket: 250mm
2 Options
Catalog number
FKCC164250
also known as FKCC/1642/50
Price (MYR)
585.00
Each
Add to cart
Standard Taper Joint:
24/40
Height (Metric):
Jacket: 250mm
Price (MYR)
585.00
Each
Add to cart
12mm I.D. indentations to support packing material
Top outer and lower inner standard taper joints of same size
Specifications
Height (Metric)
Jacket: 250mm
Type
Vacuum Jacketed Distillation Column
Standard Taper Joint
24/40