Check Order Status
Acetonitrile, UHPLC Grade, for Gradient Analysis, Fisher Chemical™
Element Search
Structure Search
Acetonitrile, UHPLC Grade, for Gradient Analysis, Fisher Chemical™
UHPLC Grade Acetonitrile for Gradient Analysis with lot-to-lot consistency that ensures reproducible results.
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCA0650PB17
also known as FKCA/0650/PB17
Price (MYR)
544.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
544.00
EA
Add to cart
Specifications
Chemical Name or Material
Acetonitrile
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Boiling Point
81°C
View more
Efficiency: lot-to-lot consistency ensures reproducible results
Low acidity/alkalinity levels avoid interactions with samples
Peak shape: low baseline noise at 210 nm and 254 nm improves peak shape
Speed: filtered to 0.1 μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity