Check Order Status
Acetonitrile, for HPLC, Fisher Chemical™
Element Search
Structure Search
Acetonitrile, for HPLC, Fisher Chemical™
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number
FKCA062617
also known as FKCA/0626/17
Price (MYR)
520.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
520.00
BOT
Add to cart
Specifications
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C