Check Order Status
Acetone, for HPLC
Element Search
Structure Search
Acetone, for HPLC
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, Amber glass bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCA060617
also known as FKCA/0606/17
Price (MYR)
223.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
223.00
EA
Add to cart
Specifications
pH
7
Melting Point
-95°C
CAS
67-64-1
Color
Colorless
Boiling Point
56°C
View more
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, Amber glass bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C