Traceable™ Dual Thermometer with Min/Max and Time/Date
Traceable™ Dual Thermometer with Min/Max and Time/Date
Actual product may vary
Fisherbrand
Traceable™ Dual Thermometer with Min/Max and Time/Date
Temperature of the two different probes is displayed in both Celsius and Fahrenheit. Traceable™ Dual Thermometer with Min/Max and Time/Date displays date and time when minimum and maximum temperatures occur.
Have Questions?
Catalog NumberProbe Type
FISS981831 Celsius, 1 Fahrenheit
Catalog number FISS98183
Price (MYR)
-
Probe Type:
1 Celsius, 1 Fahrenheit
  • Measures the temperature in the freezer and another in the refrigerator
  • Alarm type: audio and visual alarms programmable in 1° increments
  • 10 ft. micro-cable
  • Probe sealed in a miniature bottle 1 x 2.50 in. (2.5 x 6.3 cm)
  • Range: -58° to +158°F and -50° to +70°C
  • Resolution: 0.1°
  • Accuracy: ±1°C
  • Eliminates concerns about incidental contact with food or drinking water
  • High-impact, chemical-resistant ABS plastic case: 3.50 x 1.75 x 1 in. (8.9 x 4.4 x 2.5 cm)
  • Weight: 3.66 oz. (103.7 g)
  • Supplied with two special bottle probes specifically for refrigerator/freezer applications; bottle is filled with nontoxic glycol
  • Supplied with two 0.25D x 1.25 in.L (0.6 x 3.1 cm) probes
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W x H)3.5 x 1.75 x 1 in. (8.9 x 4.4 x 2.5 cm)
For Use With (Application)Cold storage, chemicals, pharmaceuticals
Height (English)1 in.
Height (Metric)2.5 cm
IncludesAAA battery for continuous, one-year always-on monitoring, flip open stand, Velcro™ and magnetic strips for mounting of both thermometer and bottle sensor, with min/max and time/date, 2 separate probes in C and F
Length (English) Cable42 in.
Length (Metric)89 mm
Length (Metric) Cable106.68 cm
MaterialABS Plastic
Probe Type1 Celsius, 1 Fahrenheit
Resolution0.1°C
Weight (English)3.6 oz.
Weight (Metric)100 g
Width (English)1.75 in.
Product TypeDigital Thermometer
Length (English)3.5 in.
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +158°F
Temperature Range (Metric)-50°C to +70°C
CalibratedYes
Battery TypeAAA
Accuracy±1°C
DisplayDigital LCD
Width (Metric)4.4 cm
ScaleCelsius, Fahrenheit
Unit SizeEA