Class A Glass Volumetric Flask with Stopper
Class A Glass Volumetric Flask with Stopper
Actual product may vary
Fisher Catalogue
Class A Glass Volumetric Flask with Stopper
Class A Glass Volumetric Flask with Stopper - Features a flat bottom design, single graduation, and blue print markings.
Have Questions?
Catalog NumberCapacity (Metric)
FISS89395100 mL
Catalog number FISS89395
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
  • Includes a solid glass stopper
  • Flat bottom design
  • Single graduation mark
  • Blue printed markings
  • Class A, in compliance with ASTM-E288
  • Autoclavable
Specifications
Capacity (English)3.38 oz.
Capacity (Metric) Media100 mL
Certifications/ComplianceASTM-E288
Closure ColorClear
Closure MaterialBorosilicate Glass 3.3
ColorClear
DescriptionGlass stopper
Diameter (Metric) Base38.1 mm
Capacity (Metric)100 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Prepare Chemical Solution
Height (English)12.5 in.
Height (Metric)16.5 cm
Length (English)5 in.
Length (Metric)5 cm
Neck StyleLong
Product TypeVolumetric Flasks
ShapeStraight-sided, Round
Tolerance±0.08 mL
Weight (English)0.2 lb.
Width (English)5 in.
Width (Metric)5 cm
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleBeaded
SterilityNon-sterile
ClassA
Bottom ShapeFlat
Closure TypeGlass Stopper
Surface TreatmentNon-treated
Closure SizeStopper No. 13
Unit SizeEA