Class B Round Base Graduated Cylinder
Class B Round Base Graduated Cylinder
Actual product may vary
Class B Round Base Graduated Cylinder
Class B Round Base Graduated Cylinder
Fisherbrand
Class B Round Base Graduated Cylinder
Eisco™ Class B Round Base Graduated Cylinder feature printed graduations and spouts.
Have Questions?
Catalog NumberCapacity (Metric)GraduationsIncludes
FISS88428100 mL1 mL1 Ea.
Catalog number FISS88428
Price (MYR)
-
Capacity (Metric):
100 mL
Graduations:
1 mL
Includes:
1 Ea.
  • Made from high quality Borosilicate 3.3 Glass
  • Single piece construction with round base
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class B
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Specifications
Base TypeRound
Description1 Ea.
Diameter (Metric) Inside29 mm
Scale ColorBlue
ShapeRound
Certifications/ComplianceDIN EN ISO 4788
ClassClass B
Height (English)9.75 in.
Height (Metric)24 cm
Tolerance Metric±1 mL
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside35 mm
Capacity (Metric)100 mL
For Use With (Application)Measuring Liquids
Includes1 Ea.
Graduations1 mL