Check Order Status
Lume Series Aspheric COB LED Lighted Hand-Held Magnifier with 7x Spot Lens and Two Brightness Settings
Actual product may vary
Lume Series Aspheric COB LED Lighted Hand-Held Magnifier with 7x Spot Lens and Two Brightness Settings
Magnifying glasses contain three ultra bright COB LED lights spread proportionally around rim of lens for wide and even lighting
Have Questions?
Catalog Number
Magnification
Diameter (Englis) Lens
FISS81723
2x
4.5 in.
Catalog number
FISS81723
Price (MYR)
-
Magnification:
2x
Diameter (Englis) Lens:
4.5 in.
For low vision tasks including reading fine print, hobby, crafts, coin and stamp inspection, model building, jewelry making
Lightweight, acrylic lenses have aspheric design
Light rays pass through lens and meet at a single point for crystal clear and distortion-free image quality
Backed by Carson™'s limited lifetime warranty
COB LED lights powered by 2 AAA batteries (not included)
Magnifiers available in two different magnifications:
2.5x Power (AS-90)
Stronger magnification but smaller lens diameter (3.5 in.)
2x power (AS-95)
Provides a wider viewing area with 4 in. lens diameter
Includes:
7x power spot lens built into handle at magnifier
Protective rim around lens to protect from scratching and breaking
Specifications
Diameter (Englis) Lens
4.5 in.
Diameter (Metric) Lens
0.114 m
Magnification
2x
Material
Acrylic
Product Type
Magnifier, Handheld Lighted