Caustic Spill Cleanup Kit
Caustic Spill Cleanup Kit
Actual product may vary
Fisherbrand
Caustic Spill Cleanup Kit
Contain, remove and tag spills fast.
Have Questions?
Catalog NumberCapacity (English)
FISS80201B0.26 gal.
Catalog number FISS80201B
Price (MYR)
-
Capacity (English):
0.26 gal.
  • One instruction sheet/SDS
  • Two gloves
  • Two 9 x 16 in. (22.9 x 40.6 cm) polybags
  • Two twist ties
  • Two blank shipping tags
  • One dust pan and brush
  • One bag vermiculite absorbent
  • One bottle neutralizing mixture (Citric Acid Anhydrous and Litmus Powder)
Specifications
ColorWhite
Capacity (English)0.26 gal.
Capacity (Metric)1 L
MaterialPolyethylene, Polypropylene
TypeChemical Spill Station
For Use With (Application)For labs
Includes1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag vermiculite absorbent, 1 bag neutralizing mixture
Packaging Quantity1 Ea.
Weight (English)5.13 lb.
Weight (Metric)2.33 kg
Unit SizeEA