Fieldmaster™ Plankton Net
Fieldmaster™ Plankton Net
Actual product may vary
Fieldmaster™ Plankton Net
Made from durable Nitex™ nylon for greater mesh size accuracy.
Have Questions?
Catalog NumberLength (English)Height (English)Diameter (English) OpeningMesh Size
FISS6811615 in.15 in.5 in.80μm
Catalog number FISS68116
Price (MYR)
-
Length (English):
15 in.
Height (English):
15 in.
Diameter (English) Opening:
5 in.
Mesh Size:
80μm
  • Greater mesh size accuracy ensures catching the correct size of plankton
  • Mini-nets are 5D x 15 in.L (12.7 x 38.1cm)
  • Student nets are 8 dia. x 20 in.L (20.3 x 50.8cm)
Specifications
Diameter (English) Opening5 in.
Diameter (Metric) Opening12.7 cm
For Use With (Application)Collecting Plankton in Shallow Water
Product TypeMini Plankton Net
Height (Metric)38.1 cm
IncludesMini plankton adaptor, one 125mL plastic bottle that attaches to the bottom of the net
Height (English)15 in.
Length (English)15 in.
MaterialNylon
Length (Metric)38.1 cm
Mesh Size80μm
Unit SizeEA