ABO-Rh Combination Blood Typing Kit
ABO-Rh Combination Blood Typing Kit
Actual product may vary
Fisherbrand
ABO-Rh Combination Blood Typing Kit
Provides information on several different subjects related to blood typing.
Have Questions?
Catalog number FISS67359
Price (MYR)
-
This popular Lab-Aids product combines all the features of Basic Blood Typing and Rh Blood Typing kits with a specially designed Slide Guide™. Provides information on the history of the blood factors, hereditary determination, individual typing techniques, comparisons of blood types in world populations, and blood typing birth problems.
  • Determine ABO blood type and Rh factor
  • Correlate agglutination to antigen production
  • Observe and study Rh mechanisms and relationships to transfusion and genetics
  • Classroom size: 32 students
Note: All sera have been tested for HBs Ag and HTLV-III (AIDS).
Specifications
Product TypeAbo-Rh Combination Blood Typing Kit
For Use With (Application)Observe and study rh mechanisms and relationships to transfusion and genetics
Includes32 Combi slide guide, one of each drop controlled bottle Blood typing Serum, type Anti-A, type Anti-B and type Anti-Rh, 32 spatulas for Anti-A Serum (blue), 32 spatulas for Anti-B Serum (yellow), 32 spatulas for Anti-Rh Serum (red), 32 pre-saturated Alcohol pads, 32 sterile disposable Blood lancets, 32 student worksheets and guides, Instruction manual with MSDS
Class Size32 Students
Unit SizeEA