Check Order Status
Glass Stirring Rods, Rounded Ends, Pack of 10
Actual product may vary
Glass Stirring Rods, Rounded Ends, Pack of 10
Glass Stirring Rods are made from high quality borosilicate glass and feature two rounded ends.
Have Questions?
Catalog Number
Length (Metric)
FISS41750
20 cm
Catalog number
FISS41750
Price (MYR)
-
Length (Metric):
20 cm
Specifications
Autoclavable
Autoclavable
Color
Clear
For Use With (Application)
Stiring
Diameter (Metric)
7 mm
Length (Metric)
20 cm
Material
Borosilicate Glass
Quantity
10
Shape
Sphere
Type
Stirring Rod