Long-Stem Digital Thermometer
Long-Stem Digital Thermometer
Actual product may vary
Fisherbrand
Long-Stem Digital Thermometer
Durable and safe, containing no mercury or alcohol. Fisherbrand Long-Stem Digital Thermometers are rugged and versatile digital thermometers to read soil, or stir and measure the temperature of solutions. Can also be used with incubators.
Have Questions?
Catalog NumberTemperature Range (Metric)
FISS407993-50°C to +150°C
Catalog number FISS407993
Price (MYR)
-
Temperature Range (Metric):
-50°C to +150°C
  • Accuracy: ±1°C
  • Large, bright 0.2 in. (6mm) display
  • Reads melting/freezing point temperatures, heat of fusion, temperatures hotter than 110°C (230°F)
  • Stainless-steel probe is resistant to acids, bases and solvents
  • Dual temperature range: -50° to +150°C (-58° to +302°F)
  • Stem length: 8 in. (20.3cm); diameter: 3mm
  • Overall length: 11 in. (28cm)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Diameter (English) Probe0.14 in.
For Use With (Application)Food, gases, samples, reagents, water baths and semi solids
IncludesStainless Steel probe traceable certificate, protective sleeve, battery
Length (English) Probe8 in.
Length (Metric)280 mm
Length (Metric) Probe20 cm
MaterialABS Plastic
Resolution0.1°C
Product TypeDigital Thermometer
Length (English)11 in.
Max. Temperature (English)302°F
Max. Temperature (Metric)150°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (Metric)-50°C to +150°C
CalibratedYes
Accuracy±1°C
DisplayDigital LCD
Width (Metric)8 cm
Unit SizeEA