Graduated HDPE Rectangular Bottles
Graduated HDPE Rectangular Bottles
Actual product may vary
Graduated HDPE Rectangular Bottles
Save valuable shelf space while providing greater materials storage. Fisherbrand™ Graduated HDPE Rectangular Bottles are available in different capacities.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (English) Inner Neck
FISS293081000 mL2.2 in.
Catalog number FISS29308
Price (MYR)
-
Capacity (Metric):
1000 mL
Diameter (English) Inner Neck:
2.2 in.
Ideal for shipping and storing solid chemicals, liquids and pastes.
  • Graduated in mL
  • Wide mouth for convenient cleaning, sample filling and retrieval
  • HDPE screw cap with LDPE inner security seal eliminates leaks
  • Attachment holes on closure and bottle shoulder permit attachment of identification tag or custody seal
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorWhite
Diameter (English) Inner Neck2.2 in.
Diameter (Metric) Inner Neck58 mm
MaterialHDPE
Length (Metric)4.14mm
QuantityCS/24, PK/6
SterilityNon-sterile
Width (Metric)3.11mm
AutoclavableNot Autoclavable
Height (Metric)176 mm
Product TypeSquare Bottle
Height (English)7 in.
Closure MaterialHDPE
Closure TypeScrew Cap
Unit SizePK