HDPE Economy Bottle Carrier
HDPE Economy Bottle Carrier
Actual product may vary
Fisherbrand
HDPE Economy Bottle Carrier
Provides economical means to transport up to six 1 Liter bottles (3.5 in. diameter or smaller).
Have Questions?
Catalog NumberHolds
FISS29066Bottle (3.5 in. dia.)
Catalog number FISS29066
Price (MYR)
-
Holds:
Bottle (3.5 in. dia.)
  • Made from HDPE
  • Lightweight and sturdy
  • Deep compartments
  • Approximate dimensions: 13 x 9.75 x 4.25 in.
  • Handle is approximately 6 in. high
Specifications
Product TypeBottle Carrier
AutoclavableNot Autoclavable
ColorGray
HoldsBottle (3.5 in. dia.)
MaterialHDPE
Positions6
Dimensions (L x W x H)13 x 9.75 x 4.25 in. (33.02 x 24.76 x 10.79cm)
Length (Metric)33.02 cm
Length (English)13 in.
Width (Metric)24.76 cm
Width (English)9.75 in.
Height (Metric)10.79 cm
Height (English)4.25 in.
Unit SizeEA