Peridotite Specimen (Igneous Rock)
Peridotite Specimen (Igneous Rock)
Actual product may vary
Peridotite Specimen (Igneous Rock)
For use in comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series. EISCO Peridotite Specimen (Igneous Rock) is iron-rich ultramafic rock, with chemistry that suggests genesis from point deep within crust.
Have Questions?
Catalog NumberQuantity
FISS24764Each
Catalog number FISS24764
Price (MYR)
-
Quantity:
Each
  • Useful as classroom demonstration of what is believed to make up most of earth's mantle, far below crust
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Contains abundant heavy elements such as iron and magnesium
  • Large crystals indicate slow cooling in magma chamber
  • Comes in clear labeled and barcoded bag for storage
Specifications
PackagingClear labeled and barcoded bag
QuantityEach
For Use With (Application)Comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series
Product TypePeridotite Study Specimen (Igneous Rock)
Width (English)Approx. 1 in.
Width (Metric)Approx. 3 cm
Unit SizeEA