Retort Clamp - 4-Prong, Powder-Coated
Retort Clamp - 4-Prong, Powder-Coated
Actual product may vary
Fisherbrand
Retort Clamp - 4-Prong, Powder-Coated
This 4 prong, cork lined retort clamp is perfect for holding lab equipment in place during delicate experiments.
Have Questions?
Catalog NumberColor
FISS24275Gray
Catalog number FISS24275
Price (MYR)
-
Color:
Gray
Specifications
AutoclavableAutoclavable
Quantity1
TypeClamp
MaterialMetal
For Use With (Equipment)Lab equipments
For Use With (Application)Apparatuses and setups
DescriptionMetal Clamps
Diameter (English)3 in.
Diameter (Metric)76.2 mm
Height (Metric)25.4 mm
Height (English)1 in.
Length (English)9.75 in.
Length (Metric)247.65 mm
Width (English)3 in.
Width (Metric)76.2 mm
ColorGray
Jaw Type4-Prong