Heavy Duty Cast Iron Retort Base
Heavy Duty Cast Iron Retort Base
Actual product may vary
Fisherbrand
Heavy Duty Cast Iron Retort Base
This base meets all international standards and is manufactured of over 4 lb. of sturdy cast iron before being power coated with a chemical resistant, visually aesthetic, blue paint.
Have Questions?
Catalog NumberDimensions (L x W)
FISS2424525 x 16 cm
Catalog number FISS24245
Price (MYR)
-
Dimensions (L x W):
25 x 16 cm
  • Rectangular retort base made of chemical-resistant cast iron with a blue coating
  • Base features a tapped hole for a 10 mm x 1.5 mm threaded rod (sold separately). Best used with Eisco Labs rod series: CH0657, CH0658, CH0659
  • The durable and sturdy construction of this set makes it an excellent choice for science classrooms and basic professional laboratory setups
Specifications
MaterialCast Iron
Height (Metric)226.79 g
Height (English)0.5 in.
Dimensions (L x W)25 x 16 cm
Length (English)10 in.
Length (Metric)25.4 cm
Width (English)6 in.
Width (Metric)15.24 cm
IncludesTapped hole 10 x 1.5 mm threaded rod
Product TypeRetort Stand Base
ShapeRectangular
Thread Size10 x 1.5 mm
AutoclavableAutoclavable
ColorBlue
DescriptionChemical-resistant
FinishPowder Coated
For Use With (Application)For science classrooms, household laboratories and basic professional laboratory setups
For Use With (Equipment)For holding and manipulating an experimental apparatus.
Quantity1
Weight (English)4.2 lb.
Weight (Metric)1.9 kg
Unit SizeEA