Borosilicate Glass Low Form Beakers
Borosilicate Glass Low Form Beakers
Actual product may vary
Borosilicate Glass Low Form Beakers
These borosilicate glass low form heavy duty beakers are general purpose media bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS11104110 mL
FISS1110405 mL
Catalog number FISS111041
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
10 mL
  • Made from low-expansion borosilicate glass.
  • Feature heavy and uniform wall thickness.
  • Spouts are designed for easy pouring.
  • Beaded tops.
  • Also include white printed graduations and a marking area.
  • Tolerance for beakers is approximately ±5%.
Specifications
Capacity (Metric)10 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)General Lab Use
Quantity12 Pk.
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
ScaleNot Graduated
StyleGriffin, Low Form
Unit SizePK