Graduated Polypropylene Pitchers with Handle
Graduated Polypropylene Pitchers with Handle
Actual product may vary
Fisherbrand
Graduated Polypropylene Pitchers with Handle
These polypropylene beakers include a handle and easy to read printed graduations.
Have Questions?
Catalog NumberCapacity (Metric)
FISS0738210 L
Catalog number FISS07382
Price (MYR)
-
Capacity (Metric):
10 L
High Quality polypropylene beakers.
  • Tapered design is convenient for stacking and gripping
  • Beakers include pour spout and easy-to-read silk-screened graduations
  • Features a comfortable rectangular handle
  • Autoclavable
  • Available in sizes ranging from 100 to 10,000 mL
Specifications
Capacity (Metric)10 L
MaterialPolypropylene
Height (Metric)330 mm
IncludesSpout
QuantityEach
TypePitcher
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
ScaleSilk Screened Print Graduated
StyleClosed Handle
Unit SizeEA