Van de Graaff Generator with Accessories
Van de Graaff Generator with Accessories
Actual product may vary
Fisherbrand
Van de Graaff Generator with Accessories
All inclusive, high-power Van De Graaff kit including 10 accessories for a variety of electrostatic demonstrations.
Have Questions?
Catalog NumberVoltage
FISS05838110 V
Catalog number FISS05838
Price (MYR)
-
Voltage:
110 V
  • Van De Graaff can generate 200,000 to 400,000 volts depending on humidity levels
  • Typical discharge arch is 2 to 8 cm
  • Precision machined dome bracket to ensure easy fit and proper dome mounting to collect charge
  • Designed for demonstrations that cover all aspects of electrostatics such as charge conservation and transfer, Triboelectric Effect, conductors, insulators, Ionization, and more
  • Kit includes: Van De Graaff, grounded base mount discharge wand (removable), dust cover, Faraday's pail, perspex cylinder, head of hair, point dis-charger, neon bulb, electric whirl, pillar with suspended metal sphere, and comb
  • Features full instruction guide for assembly, troubleshooting, pedagogy, and student activities
  • Operating voltage: 110 V
  • Size: dome diameter: 9 in.; discharge wand diameter: 4 in.; overall size: 24 in. H with 9 x 10 in. base
Specifications
For Use With (Application)Electrostatic experiments and where a continuous source of high voltage is required
Amperage∼6 μA
Voltage110 V
IncludesVan De Graaff, discharge wand, dust cover, Faraday's pail, perspex cylinder, head of hair, point dis-charger, neon bulb, electric whirl, pillar with suspended metal sphere, comb, instruction guide
Product TypeVan de Graaff Generator
Length (English)9 in.
Length (Metric)0.35 in.
Width (English)10 in.
Width (Metric)0.39 in.
Height (English)24 in.
Height (Metric)61 cm
Unit SizeEA