Check Order Status
Kimble™ Kontes™ Pellet Pestle™ Microtubes
Actual product may vary
Kimble™ Kontes™ Pellet Pestle™ Microtubes
For use with Pellet Pestle
Have Questions?
Catalog Number
Capacity (Metric)
Material
FISK749510-1501
1.5 mL
Polypropylene
Catalog number
FISK749510-1501
Price (MYR)
1,028.00
CS
Add to cart
Capacity (Metric):
1.5 mL
Material:
Polypropylene
Price (MYR)
1,028.00
CS
Add to cart
For use with Pellet Pestle
Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
Individually wrapped
Specifications
Autoclavable
Autoclavable
Color
Blue
Capacity (English)
0.05 oz.
Capacity (Metric)
1.5 mL
Material
Polypropylene
Product Type
Microtube
Dnase Free
Yes
Capacity
0.05 oz., 1.5mL