Kimble™ Kontes™ Rectangular TLC Developing Tanks
Kimble™ Kontes™ Rectangular TLC Developing Tanks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Rectangular TLC Developing Tanks
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
Have Questions?
Catalog NumberClosure TypeDepth (English)
FISK416180-0000Standard10.75 in.
Catalog number FISK416180-0000
Price (MYR)
2,415.00
EA
Add to cart
Closure Type:
Standard
Depth (English):
10.75 in.
Price (MYR)
2,415.00
EA
Add to cart
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
  • Tanks: K416180-0000, K416185-0000, K416190-0000
  • Lids: K416181-0000, K416186-0000
Specifications
Closure TypeStandard
MaterialSoda Lime Glass
TypeDeveloping Tank
Holds20 x 20 cm or four 5 x 20 cm Plates
ShapeRectangular
Dimensions (L x W)4 x 11.75 in.
Length (English)4 in.
Length (Metric)10.16 cm
Width (English)11.75 in.
Width (Metric)29.8 cm
Depth (English)10.75 in.
Depth (Metric)27.3 cm
Unit SizeEA