Kimble™ Plain Reusable Centrifuge Tube with Screw Caps
Kimble™ Plain Reusable Centrifuge Tube with Screw Caps
Actual product may vary
Fisher Catalogue
Kimble™ Plain Reusable Centrifuge Tube with Screw Caps
With PTFE/rubber lined black phenolic screw cap
Have Questions?
Catalog NumberCapacity (Metric)
FISK410090-001515 mL
Catalog number FISK410090-0015
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
15 mL
With PTFE/rubber lined black phenolic screw cap
  • Constructed of 33 Expansion, low-extractable borosilicate glass
  • Economical, one-piece construction
  • Autoclavable
Specifications
Certifications/ComplianceUSP, ASTM E 438
Closure MaterialPhenolic
Capacity (Metric)15 mL
MaterialBorosilicate Glass
Liner MaterialPTFE, Rubber
Thread Size15-415
AutoclavableAutoclavable
ShapeConical
Length (Metric)130 mm
Bottom ShapeConical
Diameter (Metric)17 mm
Product TypeCentrifuge Tube
Max. RCF2980 x g
Unit SizeCS