Kimble™ Kontes™ Reaction-Jacketed Beakers
Kimble™ Kontes™ Reaction-Jacketed Beakers
Actual product may vary
Kimble™ Kontes™ Reaction-Jacketed Beakers
Kimble™ Kontes™ Reaction-Jacketed Beakers
Fisher Catalogue
Kimble™ Kontes™ Reaction-Jacketed Beakers
The open top provides easy access to concentrated residual samples.
Have Questions?
Catalog NumberCapacity (Metric)
FISK317000-0250250 mL
Catalog number FISK317000-0250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
The open top provides easy access to concentrated residual samples.
  • A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
  • Jacket has serrated 3/8 in. hose connections
  • Design meets ASTM Specification E960, Type I requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
DescriptionConnection: 0.37 in. Serrated Hose
Diameter (English) Inner2.55 in.
Diameter (English) Outer3.54 in.
Diameter (Metric) Inner6.5 cm
Diameter (Metric) Outer9 cm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
For Use With (Application)Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions
Height (English) Inner3.3 in.
Height (English) Outer3.74 in.
Height (Metric) Inner8.4 cm
Height (Metric) Outer9.5 cm
IncludesConical Lower Interior Surface
TypeBeaker
Certifications/ComplianceASTM E960, ASTM E438, USP Type I
StyleOpen Top
Unit SizeEA