Sterile PETG Media Bottles
Sterile PETG Media Bottles
Actual product may vary
Sterile PETG Media Bottles
Sterile PETG Media Bottles
Sterile PETG Media Bottles
Fisherbrand
Sterile PETG Media Bottles
Reduce the risk of cross contamination with these sterile, disposable media bottles
Have Questions?
Catalog NumberCapacity (Metric)Closure Type
FISFBMB3030 mLTamper-evident Screw Cap
Catalog number FISFBMB30
Price (MYR)
1,306.00
CS
Add to cart
Capacity (Metric):
30 mL
Closure Type:
Tamper-evident Screw Cap
Price (MYR)
1,306.00
CS
Add to cart
Reduce the risk of cross contamination with these sterile, disposable media bottles
The Fisherbrand PETG Media Bottle is ideal for media preparation and storage.
  • An inexpensive and shatterproof alternative to glass media bottles
  • Nonpyrogenic and noncytotoxic to protect cells
  • Molded-in graduations won't rub off
  • Made from clear, lightweight, polyethylene terephthalate copolyester (PETG) providing impact resistance and excellent gas-barrier properties
  • Closure is is high density polyethylene (HDPE)
  • Bottles packed in a shrink-wrapped tray to stay clean
  • Heat-shrink band around cap and bottle neck provides visual assurance of sterility for individual bottles until band is removed
  • Part numbers ending in "A" are designed without a temper-evident ring
Specifications
Capacity1.0 oz., 30mL
Capacity (Metric)30 mL
Capacity (English)1.0 oz.
ColorClear
For Use With (Application)For media preparation and storage
MaterialPETG
SterilitySterile
Certifications/ComplianceSAL 10-6, USP Class VI
Product TypeMedia Bottle
Closure MaterialHDPE
Closure Size20 mm
Closure TypeTamper-evident Screw Cap
Unit SizeCS