Check Order Status
Porcelain Rose Crucible
Actual product may vary
Porcelain Rose Crucible
For use in very high temperatures
Have Questions?
Catalog Number
Capacity (Metric)
FISFB960Z
75 mL
Catalog number
FISFB960Z
Price (MYR)
160.00
PK
Add to cart
Capacity (Metric):
75 mL
Price (MYR)
160.00
PK
Add to cart
Unglazed
Without perforated cover or tube
Color: white
Capacity: 2.54 oz. (75mL)
Specifications
Material
Porcelain
For Use With (Application)
Used in very high temperature
Capacity (Metric)
75 mL
Product Type
Rose Crucible