Reusable Test Tubes with Rubber-Lined Screw Caps
Reusable Test Tubes with Rubber-Lined Screw Caps
Actual product may vary
Fisherbrand
Reusable Test Tubes with Rubber-Lined Screw Caps
25 x 150mm
Have Questions?
Catalog NumberDiameter (Metric)
FISFB7012515025 mm
Catalog number FISFB70125150
Price (MYR)
1,028.00
PK
Add to cart
Diameter (Metric):
25 mm
Price (MYR)
1,028.00
PK
Add to cart
Black phenolic cap with inert rubber liner
  • White marking spot
  • GPI Thread Closure 24-410
Specifications
Certifications/ComplianceASTM E982, Type VI, Class A
Closure MaterialPhenolic
Capacity (Metric)50 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Liner MaterialRubber
Thread Size24-410
SterilityNon-sterile
Diameter (Metric)25 mm
Closure TypeScrew Cap
Product TypeReusable Culture Tube
Unit SizePK