Round-Bottom Polystyrene Test Tubes With Cap
Round-Bottom Polystyrene Test Tubes With Cap
Actual product may vary
Round-Bottom Polystyrene Test Tubes With Cap
Round-Bottom Polystyrene Test Tubes With Cap
Round-Bottom Polystyrene Test Tubes With Cap
Round-Bottom Polystyrene Test Tubes With Cap
Round-Bottom Polystyrene Test Tubes With Cap
Fisherbrand™ round-bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
Have Questions?
Catalog NumberCapacity (Metric)SterilityClosure TypePackaging
FISFB149564A8 mLSterileScrew CapBlister Pk.
Catalog number FISFB149564A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
8 mL
Sterility:
Sterile
Closure Type:
Screw Cap
Packaging:
Blister Pk.
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
ColorClear
Capacity (Metric)8 mL
Length (Metric)100 mm
MaterialPolystyrene
DisposableSingle-use
PackagingBlister Pk.
SterilitySterile
Product LineFisherbrand
GraduatedNo
Closure TypeScrew Cap
Product TypeTest Tube
Unit SizeCS