Straight Sided Jars with White Polypropylene Caps
Straight Sided Jars with White Polypropylene Caps
Actual product may vary
Straight Sided Jars with White Polypropylene Caps
Fisherbrand™ Straight Sided Jars with White Polypropylene Caps are ideal for large solid samples and allow for complete removal of contents.
Have Questions?
Catalog NumberCapacity (Metric)
FISFB02912026A60 mL
Catalog number FISFB02912026A
Price (MYR)
188.00
CS
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
188.00
CS
Add to cart
  • Translucent and durable
  • Jars are ideal for storing and collecting specimens and lab supplies
  • Wide neck and straight sides facilitate easy recovery of contents
  • Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity
  • Features wide neck and straight sides to facilitate easy recovery of contents
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorNatural
Diameter (Metric) Outer Neck50 mm
Diameter (Metric) Inner Neck47 mm
For Use With (Application)Large Solid Samples and Allow for Complete Removal of Contents
IncludesCap
Height (Metric) With Closure48 mm
MaterialPolypropylene
Quantity48
Closure ColorWhite
Neck Finish53-400
ClarityTranslucent
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)45 mm
Product TypeStraight-sided Jar
Closure MaterialPolypropylene
Closure Size53-400 mm
Diameter (Metric)48 mm
Closure TypeCap
Unit SizeCS