Amber Boston Round Glass Bottles with Black Phenolic Caps
Amber Boston Round Glass Bottles with Black Phenolic Caps
Actual product may vary
Amber Boston Round Glass Bottles with Black Phenolic Caps
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure LinerQuantity
FISFB02911900240 mLPolyethylene Cone108
FISFB02911735120 mLPolyethylene Cone24
FISFB02911743480 mLPolyethylene Cone12
Catalog number FISFB02911900
Price (MYR)
877.00
CS
Add to cart
Capacity (Metric):
240 mL
Closure Liner:
Polyethylene Cone
Quantity:
108
Price (MYR)
877.00
CS
Add to cart
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
  • Narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorbs all wavelengths in the ultraviolet spectrum
  • Black Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier
  • Liners mold themselves around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Ideal for everything from liquids, to soaps, to macro-invertebrates.
Specifications
Capacity (Metric)240 mL
Capacity (English)8 oz.
ColorAmber
Diameter (Metric) Outer Neck21 mm
Diameter (Metric) Inner Neck13 mm
Height (Metric) With Closure139 mm
MaterialGlass (Type 3)
Quantity108
Closure ColorBlack
Neck Finish24-400
Closure IncludedYes
MouthNarrow
Height (Metric)136 mm
Product TypeBoston Round Bottle
Closure LinerPolyethylene Cone
Closure MaterialPhenolic
Closure Size24-400 mm
Diameter (Metric)60 mm
Unit SizeCS