Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Actual product may vary
Fisher Catalogue
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Amber, USP Type III soda-lime glass ideal for storage of light sensitive samples or dry powders. Easy removal of contents.
Have Questions?
Catalog NumberCapacity (Metric)Closure Liner
FISFB02911441250 mLPTFE/Polyethylene Foam
Catalog number FISFB02911441
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Liner:
PTFE/Polyethylene Foam
  • Wide opening for easy transfer of contents
  • Amber color filters out ultraviolet wavelengths, ideal for light sensitive solids
  • Classic shape of vitamin bottles, often used in pharmaceutical packaging
  • Often used in chemical and environmental applications
  • Caps offer good impact strength and pliability, can be remelted after they are formed
  • Cap liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
Capacity (Metric)250 mL
Capacity (English)8.5 oz.
ColorAmber
Diameter (Metric) Outer Neck41 mm
Diameter (Metric) Inner Neck32 mm
Height (Metric) With Closure122 mm
MaterialGlass (Type 3)
Quantity24
Closure ColorWhite
Neck Finish45-400
Closure IncludedYes
MouthWide
Height (Metric)119 mm
Product TypeWide Mouth Packer Bottle
Closure LinerPTFE/Polyethylene Foam
Closure MaterialPolypropylene
Closure Size45-400 mm
Diameter (Metric)65 mm
Unit SizeCS