Plastilab™ All-Purpose Cylindrical Stirring Bars
Plastilab™ All-Purpose Cylindrical Stirring Bars
Actual product may vary
Fisher Catalogue
Plastilab™ All-Purpose Cylindrical Stirring Bars
Features rounded shape with smooth walls.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FIS75-011.68 mm4.66 mm
Catalog number FIS75-0
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
11.68 mm
Diameter (Metric):
4.66 mm
Features rounded shape with smooth walls.
  • PTFE-coated magnet
  • Magnetic field causes stirring bar to spin very quickly
  • Non-stick, non-reactive and hydrophobic
  • Highly resistant to thermo-shock
  • Suitable for foodstuff
Specifications
DescriptionMagnet PTFE Coated
Diameter (Metric)4.66 mm
Length (Metric)11.68 mm
Quantity10/Pk.
ShapeCylinder, Sphere
Unit SizeEA