Wood Handled Cotton Swabs and Applicators
Actual product may vary
Wood Handled Cotton Swabs and Applicators
Wood Handled Cotton Swabs and Applicators
Wood Handled Cotton Swabs and Applicators
Fisherbrand
Wood Handled Cotton Swabs and Applicators
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)QuantitySterilityProduct Type
FIS22-363-1576 in.1000/pkNon-sterileCotton Tip Applicator
FIS22-363-1736 in.1/ envelope; 1000/csSterileCotton Tip Applicator
Catalog number FIS22-363-157
Price (MYR)
27.00
Pack
Length (English):
6 in.
Quantity:
1000/pk
Sterility:
Non-sterile
Product Type:
Cotton Tip Applicator
Price (MYR)
27.00
Pack
  • Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
  • Sterile and non-sterile products available (see product specifications)
  • Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
  • Not suitable for PCR DNA/ RNA testing
Specifications
AutoclavableAutoclavable
For Use With (Application)Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments
Length (English)6 in.
Length (Metric)15 cm
Quantity1000/pk
SterilityNon-sterile
Handle MaterialWood
Product TypeCotton Tip Applicator
Tip MaterialCotton
Unit SizePack