Slide Holder
Slide Holder
Actual product may vary
Slide Holder
Twelve-place files keep slides separated, visible and orderly near the microscope in use
Have Questions?
Catalog number FIS22-261926
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Made from vacuum-formed polystyrene
  • Allows slides to project over the rack edge for easy handling
  • Dimensions 8.6 x 17.8 x 1.9cm H (3.375 x 7 x 0.75 in. H)
Specifications
TypeSlide Holder
Slide Capacity12
ColorWhite
Device Holds12
Dimensions (L x W x H)7 x 7 x 0.75 in. (17.8 x 17.8 x 1.9 cm)
For Use With (Equipment)Microscope Slide
Height (English)0.75 in.
Height (Metric)1.9 cm
Length (English)7 in.
Length (Metric)17.8 cm
MaterialPolystyrene
Width (English)7 in.
Width (Metric)17.8 cm
Unit SizePK