Smooth, chemical-resistant scrapers useful for removing residues from filters, cleaning containers, mixing and distributing solids in fluids, spreading pastes over large areas
Have Questions?
Catalog Number
Length (Metric) Handle
Length (English) Handle
FIS22-261596
183 cm
72 in.
Catalog number FIS22-261596
Price (MYR)
2,113.00
EA
Add to cart
Length (Metric) Handle:
183 cm
Length (English) Handle:
72 in.
Price (MYR)
2,113.00
EA
Add to cart
Molded from HDPE plastic
Blade tapers from 9.5 mm (0.375 in.) down to a chisel edge
Upper end extends into a hexagonal shank 35 mm (1.375 in.) wide