Check Order Status
Disposable Wintrobe Tube
Actual product may vary
Disposable Wintrobe Tube
Labelled with white enamel for easy reading
Have Questions?
Catalog Number
Quantity
FIS22-150730
Each
Catalog number
FIS22-150730
Price (MYR)
393.00
PK
Add to cart
Quantity:
Each
Price (MYR)
393.00
PK
Add to cart
Labelled with white enamel for easy reading
Precision-scaled from the interior base
Ideal for determining sedimentation rates and hematocrit values
Specifications
CE Marker
No
Type
Wintrobe Tube