LeStreak™ Microstreakers
LeStreak™ Microstreakers
Actual product may vary
LeStreak™ Microstreakers
Perfect for inoculating or transferring microbiological samples
Have Questions?
Catalog NumberVolumeCalibratedDiameter (Metric) Inside
FIS22-0320965.45 μLNo0.35 cm
Catalog number FIS22-032096
Price (MYR)
-
Volume:
5.45 μL
Calibrated:
No
Diameter (Metric) Inside:
0.35 cm
Perfect for inoculating or transferring microbiological samples
  • Use to inoculate or transfer microbiological samples
  • Made of high-quality twisted nichrome wire permanently attached to lightweight insulated aluminum handles
  • Slide smoothly across agar surfaces without tearing or gouging
  • Nichrome wire cools rapidly after flaming
  • 1 and 10μL loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test
  • Noncalibrated microstreakers are color coded for easy identification
Specifications
Diameter (English) Inside0.14 in.
Diameter (Metric) Inside0.35 cm
Length (English)10.75 in.
Length (Metric)27.31 cm
MaterialNichrome
SterilityNon-sterile
Volume5.45 μL
SizeLarge
ColorRed
Product TypeInoculating Streaker
IncludesNichrome wire loop permanently attached to red insulated aluminum holder
CalibratedNo
Unit SizePK