Check Order Status
Fit Test Kit Accessory, Smoke Tube
Actual product may vary
Fisher Catalogue
Fit Test Kit Accessory, Smoke Tube
Allegro™ Fit Test Kit Accessory, Smoke Tube is used for qualitative fit testing of air purifying respirators.
Have Questions?
Catalog number
FIS19-083018
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Replacement tubes
For qualitative fit testing of air purifying respirators
Specifications
Product Type
Smoke Tube
Certifications/Compliance
OSHA
For Use With (Application)
For qualitative fit testing of air purifying respirators
For Use With (Equipment)
Allegro fit test kit
Model
2050-01
Quantity
6/Pk.