AlphaTec™ 56-910 PVC Coat Apron
AlphaTec™ 56-910 PVC Coat Apron
Actual product may vary
AlphaTec™ 56-910 PVC Coat Apron
Medium to heavy duty splash protection coat aprons offers greater body coverage.
Have Questions?
Catalog number FIS19-010-247
Price (MYR)
2,776.00
CS
Add to cart
Price (MYR)
2,776.00
CS
Add to cart
  • Reliable protection against oil, grease, fats, chemicals, abrasion, punctures
  • No exposed elastic or metal parts to cause static or corrosion
  • Protection where splashes from acids and caustic bases are a concern
  • Virgin vinyl construction contains no reprocessed materials and is flexible
  • Sleeves come with elasticized cuffs for added protection
  • PVC (vinyl) grommets are permanently sealed to the aprons for durability and allow for size adjustment without tear-out
  • Adjustable ties ensure optimal fit for a wide range of wearers
N/A
Specifications
TypeApron
SizeUniversal
MaterialVinyl
ColorBlue
DisposableReusable
Closure TypeTies
SterilityNon-sterile
ConstructionVirgin Vinyl, No Reprocessed Materials, Dielectrically Sealed
FinishDielectrically Sealed
Latex FreeNo
No. of PocketsNone
ResistancePuncture, Abrasion, Chemicals, Fats, Oils, Grease
Thickness8 mil
For Use With (Application)Food Service, Maintenance, Transferring Liquids
No. per Case12/Pk.
No. per Pack1 Ea.
Seam TypeSeamless
Unit SizeCS