Check Order Status
Filter Disc, Untreated, Ground Cylindrical, 90 mm
Actual product may vary
Filter Disc, Untreated, Ground Cylindrical, 90 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Have Questions?
Catalog Number
Porosity
FIS15-909
00
Catalog number
FIS15-909
Price (MYR)
392.16
Special offer
392.16
EA
Add to cart
Porosity:
00
Price (MYR)
392.16
Special offer
392.16
EA
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
No. per Pack
5/Pk.
Porosity
00
Thickness (Metric)
6.5 ± 1.5 mm
Diameter (Metric)
90 mm