Check Order Status
Disposable Flint Glass Tubes with Plain End
Actual product may vary
Disposable Flint Glass Tubes with Plain End
Specially annealed to reduce breakage along stress point
Have Questions?
Catalog Number
Capacity (Metric)
Length (Metric)
Diameter (Metric)
Quantity
FIS14-958G
19 mL
125 mm
16 mm
250/Tray
Catalog number
FIS14-958G
Price (MYR)
728.00
CS
Add to cart
Capacity (Metric):
19 mL
Length (Metric):
125 mm
Diameter (Metric):
16 mm
Quantity:
250/Tray
Price (MYR)
728.00
CS
Add to cart
Specially annealed to reduce breakage along stress point
Round bottom
Smooth top and bottom to prevent cuts
Specifications
Capacity (Metric)
19 mL
Length (Metric)
125 mm
Material
Flint Glass
Disposable
Single-use
For Use With
Specially annealed to reduce breakage along stress point
Label Type
None
Packaging
Tray
Quantity
250/Tray
Sterility
Non-sterile
Diameter (Metric)
16 mm
Closure Type
KIM-KAP
Product Type
Disposable Culture Tube