Magnetic Stir Bar Six Packs
Actual product may vary
Fisherbrand
Magnetic Stir Bar Six Packs
These magnetic stirring bars are the most commonly used shape.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS14-513-51SIX25.4 mm9.5 mm1 in.0.375 in.
FIS14-513-57SIX12 mm4.5 mm0.44 in.0.19 in.
FIS14-513-58SIX12.7 mm8 mm0.5 in.0.31 in.
FIS14-513-59SIX25.4 mm8 mm1 in.0.31 in.
FIS14-513-60SIX38.1 mm8 mm1.5 in.0.31 in.
Catalog number FIS14-513-51SIX
Price (MYR)
64.00
Pack
Length (Metric):
25.4 mm
Diameter (Metric):
9.5 mm
Length (English):
1 in.
Diameter (English):
0.375 in.
Price (MYR)
64.00
Pack
These magnetic stirring bars are the most commonly used shape.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Specifications
AutoclavableAutoclavable
ColorWhite
Diameter (English)0.375 in.
Length (English)1 in.
CoatingPTFE
Diameter (Metric)9.5 mm
Length (Metric)25.4 mm
MaterialAlnico Magnet
Quantity6 Pk.
ShapeCylinder
TypeMagnetic Stir Bar Set
Unit SizePack