Safety Stand for Overhead Stirrers
Safety Stand for Overhead Stirrers
Actual product may vary
Safety Stand for Overhead Stirrers
Safety Stand for Overhead Stirrers
Fisherbrand
Safety Stand for Overhead Stirrers
Safety stand style for easy mounting of overhead stirrers
Have Questions?
Catalog NumberLength (Metric)
FIS14-500-354711 mm
Catalog number FIS14-500-354
Price (MYR)
3,021.00
Each
Add to cart
Length (Metric):
711 mm
Price (MYR)
3,021.00
Each
Add to cart
  • Includes 28 in. rod, stand base and allen key
  • Cast zinc-aluminum alloy base coated with chemical resistant epoxy paint
  • Two level adjustment knobs
  • Rod is 304 stainless steel tube, 1 in. diameter
  • Weight: 5 kg (11 lb.)
Specifications
Diameter (English) Shaft1 in.
Diameter (Metric) Shaft25 mm
For Use With (Equipment)Caframo™ ReversingDigital™ Overhead Stirrer
For Use With (Application)For easy mounting of overhead stirrer
IncludesTwo level adjustment knobs, 27.5 L x 1 in. dia. stainless steel tube rod, stand base and allen key
Length (English)28 in.
Length (Metric)711 mm
TypeSafety Lab Stand
Width (English)16.375 in.
Width (Metric)416 mm
Weight (Metric)5 kg
Dimensions (L x W x H)430 x 416 x 711 mm (16.4 x 16.9 x 28 in.)
CoatingChemical-resistant Epoxy Paint
MaterialCast Zinc, Aluminum
Weight (English)11 lb.
Unit SizeEach