Sterileware™ Sterile Styrene Sampler Spoons
Actual product may vary
Fisherbrand
Sterileware™ Sterile Styrene Sampler Spoons
Polystyrene plastic sterile sampler spoons are designed to gather small amounts of sample material.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FIS14-429B2.46 mL200/Cs.
Catalog number FIS14-429B
Price (MYR)
2,554.00
Case
Capacity (Metric):
2.46 mL
Quantity:
200/Cs.
Price (MYR)
2,554.00
Case
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile, individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8 cm (7 in.)
Specifications
Quantity200/Cs.
SterilitySterile
AutoclavableNot Autoclavable
Capacity (Metric)2.46 mL
ColorWhite
Certifications/ComplianceFDA
DisposableSingle-use
Product TypeSpoon
For Use With (Application)Designed to gather small amounts of sample material
Length (English)7 in.
Width (English)0.98 in.
Width (Metric)2.5 cm
Dimensions (L x W)17.8 x 2.5 cm
IncludesCertificate of sterilization
MaterialPolystyrene
Handle TypeLong
Length (Metric)17.8 cm
Unit SizeCase