Check Order Status
Solid Rubber Stoppers
Actual product may vary
Fisher Catalogue
Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
Have Questions?
Change view
Catalog Number
Length (Metric)
Size
Stopper Number
FIS14-130P
2.5 cm
10
10
FIS14-130M
2.5 cm
8
8
2 Options
Catalog number
FIS14-130P
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
2.5 cm
Size:
10
Stopper Number:
10
Available in a variety of sizes ranging from 000 through 16
Solid rubber construction
Specifications
For Use With (Application)
General purpose use
Length (Metric)
2.5 cm
Size
10
Color
Black
Diameter (Metric) Bottom
4.2 cm
Diameter (Metric) Top
5 cm
Material
Natural Rubber
Product Type
Solid Stopper
Stopper Number
10