Disposable Borosilicate Glass Pasteur Pipets
Actual product may vary
Fisherbrand
Disposable Borosilicate Glass Pasteur Pipets
Constriction near top prevents plugs from slipping into pipette body
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Quantity
FIS13-678-20B14.6 cm1440/Cs. (4 boxes of 360)
FIS13-678-20A14.6 cm720/Cs.
FIS13-678-20C22.86 cm720/Cs.
FIS13-678-20D22.86 cm1440/Cs.
Catalog number FIS13-678-20B
Price (MYR)
627.00
Case
Length (Metric):
14.6 cm
Quantity:
1440/Cs. (4 boxes of 360)
Price (MYR)
627.00
Case
High-quality pipets with square cut ends and constriction near top
  • High-quality borosilicate glass
  • Plugs not supplied
  • Tips held to close diameter tolerances
  • Square cut end
Specifications
Certifications/ComplianceASTM E732
MaterialBorosilicate Glass
IncludesSquare cut ends, constriction near top
Length (English)5.75 in.
Quantity1440/Cs. (4 boxes of 360)
Volume (Metric)2 mL
Length (Metric)14.6 cm
TypePasteur Pipet