Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Actual product may vary
Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Rugged construction
Have Questions?
Catalog NumberVolume (Metric)ColorTolerance
FIS13-664E2 mLGreen±0.02 mL
Catalog number FIS13-664E
Price (MYR)
1,720.00
CS
Add to cart
Volume (Metric):
2 mL
Color:
Green
Tolerance:
±0.02 mL
Price (MYR)
1,720.00
CS
Add to cart
Rugged construction
  • Intended for chemical laboratory work—tip openings are smaller than usually desired for clinical laboratory operations
  • Permanent brown, graduated scale fused into uniform bore tubing ending above taper
  • Color coded by size for ease in sorting
  • Tempered tips
  • Calibrated “To Deliver”
Specifications
CalibrationTo Deliver
Certifications/ComplianceASTM E438, USP Type I, Class B
ClassB
MaterialBorosilicate Glass
GraduatedYes
Graduations0.1 mL
Length (English)13.7 in.
Tolerance±0.02 mL
Volume (English)0.06 oz.
ColorGreen
Volume (Metric)2 mL
Length (Metric)350 mm
TypeGraduated Pipet, Type 1