Reusable Class B Volumetric Pipets
Reusable Class B Volumetric Pipets
Actual product may vary
Fisherbrand
Reusable Class B Volumetric Pipets
Ensure the highest accuracy and reproducibility.
Have Questions?
Catalog NumberVolume (Metric)Min. Flow TimeAccuracy
FIS13-650N20 mL10 sec.±0.06 mL
Catalog number FIS13-650N
Price (MYR)
117.00
Each
Add to cart
Volume (Metric):
20 mL
Min. Flow Time:
10 sec.
Accuracy:
±0.06 mL
Price (MYR)
117.00
Each
Add to cart
Ensure the highest accuracy and reproducibility.
  • Borosilicate glass
  • Permanent amber markings
  • Large numeral on bulb of 15mL and larger pipets makes capacity easy to identify
  • Accuracy printed on each pipet
  • Tuf-Temp™–treated tempered tips reduce chipping
  • Fast outflow
Specifications
Accuracy±0.06 mL
CalibrationTo Deliver
Certifications/ComplianceASTM E969-95
ClassB
MaterialBorosilicate Glass
GraduatedYes
Graduation ColorAmber
Min. Flow Time10 sec.
Tip StyleTuf-Temp™ treated
Volume (English)0.68 oz.
ColorYellow
Volume (Metric)20 mL
TypeVolumetric Pipet